Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240052

Sigma-Aldrich

3-(Dimethylamino)-1-propylamine

≥99%

Synonym(s):

N,N-Dimethyl-1,3-diaminopropane, N,N-Dimethyl-1,3-propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2N(CH2)3NH2
CAS Number:
Molecular Weight:
102.18
Beilstein/REAXYS Number:
605293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.6 (vs air)

vapor pressure

5 mmHg ( 20 °C)

assay

≥99%

refractive index

n20/D 1.435 (lit.)

bp

133 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCCN

InChI

1S/C5H14N2/c1-7(2)5-3-4-6/h3-6H2,1-2H3

InChI key

IUNMPGNGSSIWFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

89.6 °F - closed cup

flash_point_c

32 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yusuke Yamamoto et al.
The Journal of toxicological sciences, 44(9), 585-600 (2019-09-03)
Amino acid derivative reactivity assay (ADRA) has previously been developed as an alternative method to direct peptide reactivity assay (DPRA) to evaluate key event 1 in skin sensitization mechanisms. However, when using alternative methods for skin sensitization, integrated approaches to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service