Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239771

Sigma-Aldrich

Ethylene glycol diacetate

≥99%

Synonym(s):

1,2-Diacetoxyethane, Ethylene diacetate, Glycol diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OCOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein/REAXYS Number:
1762308
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

5.04 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

assay

≥99%

autoignition temp.

899 °F

expl. lim.

1.6 %, 135 °F
8.4 %, 154 °F

refractive index

n20/D 1.415 (lit.)

bp

186-187 °C (lit.)

mp

−41 °C (lit.)

density

1.104 g/mL at 20 °C (lit.)

SMILES string

CC(=O)OCCOC(C)=O

InChI

1S/C6H10O4/c1-5(7)9-3-4-10-6(2)8/h3-4H2,1-2H3

InChI key

JTXMVXSTHSMVQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service