Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

235164

Sigma-Aldrich

Tributyltin cyanide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnCN
CAS Number:
Molecular Weight:
316.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

mp

105-107 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)C#N

InChI

1S/3C4H9.CN.Sn/c3*1-3-4-2;1-2;/h3*1,3-4H2,2H3;;

Inchi Key

GMYAQHAKWKXYHG-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service