233374
Pyridine-d5
99 atom % D, contains 1 % (v/v) TMS
Synonym(s):
Pentadeuteropyridine
About This Item
isotopic purity
99 atom % D
assay
99% (CP)
contains
1 % (v/v) TMS
expl. lim.
0.34-6.3 % (lit.)
refractive index
n20/D 1.506 (lit.)
bp
114.4 °C (lit.)
mp
-41 °C (lit.)
density
1.05 g/mL at 25 °C (lit.)
mass shift
M+5
SMILES string
[2H]c1nc([2H])c([2H])c([2H])c1[2H]
InChI
1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D
InChI key
JUJWROOIHBZHMG-RALIUCGRSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
62.6 °F
flash_point_c
17 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service