Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

230642

Sigma-Aldrich

4-Heptylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6C6H4CO2H
CAS Number:
Molecular Weight:
220.31
Beilstein/REAXYS Number:
2047135
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid crystal

SMILES string

CCCCCCCc1ccc(cc1)C(O)=O

InChI

1S/C14H20O2/c1-2-3-4-5-6-7-12-8-10-13(11-9-12)14(15)16/h8-11H,2-7H2,1H3,(H,15,16)

InChI key

VSUKEWPHURLYTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Maria Moreno et al.
Materials (Basel, Switzerland), 12(12) (2019-06-20)
Different metalorganic lamellar hybrid materials based on associated nanoribbons were synthesized by the use of alkyl-benzyl monocarboxylate spacers, containing alkyl tails with variable lengths, which acted like structural growing inhibitors. These molecular agents were perpendicularly located and coordinated to aluminium
Hyunshun Shin et al.
Bioorganic & medicinal chemistry, 15(7), 2617-2623 (2007-02-14)
The first committed step in lipid A biosynthesis is catalyzed by uridine diphosphate-(3-O-(R-3-hydroxymyristoyl))-N-acetylglucosamine deacetylase (LpxC), a zinc-dependent deacetylase, and inhibitors of LpxC may be useful in the development of antibacterial agents targeting a broad spectrum of Gram-negative bacteria. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service