Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

230383

Sigma-Aldrich

Cobalt(II) sulfate hydrate

Synonym(s):

Cobaltous sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoSO4 · xH2O
CAS Number:
Molecular Weight:
155.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302

composition

Degree of hydration, 6-7

density

2.03 g/mL at 25 °C (lit.)

SMILES string

O.[Co++].[O-]S([O-])(=O)=O

InChI

1S/Co.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

BGORGFZEVHFAQU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service