229105
Malachite Green Carbinol base
Dye content 90 %
Synonym(s):
Solvent Green 1
About This Item
Recommended Products
composition
Dye content, 90%
mp
112-114 °C (lit.)
λmax
431 nm
SMILES string
CN(C)c1ccc(cc1)C(O)(c2ccccc2)c3ccc(cc3)N(C)C
InChI
1S/C23H26N2O/c1-24(2)21-14-10-19(11-15-21)23(26,18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,26H,1-4H3
InChI key
LXHOGENDFZKPSF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service