Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

227145

Sigma-Aldrich

1,1-Dichloro-2-propanone

>96%

Synonym(s):

1,1-Dichloroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCHCl2
CAS Number:
Molecular Weight:
126.97
Beilstein/REAXYS Number:
1740214
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

>96%

refractive index

n20/D 1.446 (lit.)

bp

117-118 °C (lit.)

density

1.327 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C(Cl)Cl

InChI

1S/C3H4Cl2O/c1-2(6)3(4)5/h3H,1H3

InChI key

CSVFWMMPUJDVKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

114.4 °F - closed cup

flash_point_c

45.8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service