Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225975

Sigma-Aldrich

Phenylmercuric nitrate, basic

Synonym(s):

Phenylmercury nitrate, Phenylmercuric nitrate, basic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5HgNO3·C6H5HgOH
CAS Number:
Molecular Weight:
634.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

175-185 °C (dec.) (lit.)

SMILES string

O[Hg]c1ccccc1.[O-][N+](=O)O[Hg]c2ccccc2

InChI

1S/2C6H5.2Hg.NO3.H2O/c2*1-2-4-6-5-3-1;;;2-1(3)4;/h2*1-5H;;;;1H2/q;;2*+1;-1;/p-1

InChI key

DQZCXZOUXZTNCU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service