Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225525

Sigma-Aldrich

2-Amino-2-norbornanecarboxylic acid

98%

Synonym(s):

2-Aminobicyclo[2.2.1]heptane-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

mp

>300 °C (lit.)

SMILES string

NC1(C[C@@H]2CC[C@H]1C2)C(O)=O

InChI

1S/C8H13NO2/c9-8(7(10)11)4-5-1-2-6(8)3-5/h5-6H,1-4,9H2,(H,10,11)/t5-,6+,8?/m1/s1

InChI key

MPUVBVXDFRDIPT-RSHNMJPRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Inhibitor of amino acid transport across cell membranes.[1][2][3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwini L Chand et al.
Human reproduction (Oxford, England), 26(11), 3102-3108 (2011-09-15)
Little is known about metabolic processes in the developing ovarian follicle. Using mouse ovarian follicles, we investigated uptake of L-leucine by follicles at varying stages of maturity in the presence of insulin-like growth factor (IGF)-1. METHODS Mouse ovarian follicles were
Laura J Brown et al.
The Journal of biological chemistry, 284(51), 35359-35367 (2009-10-28)
The cytosolic malic enzyme (ME1) has been suggested to augment insulin secretion via the malate-pyruvate and/or citrate-pyruvate shuttles, through the production of NADPH or other metabolites. We used selectable vectors expressing short hairpin RNA (shRNA) to stably decrease Me1 mRNA
Jordan A Torok et al.
American journal of respiratory cell and molecular biology, 47(1), 37-43 (2012-02-11)
Inhaled nitric oxide (iNO) is used to treat pulmonary hypertension and is being investigated for prevention of bronchopulmonary dysplasia in neonates. Extrapulmonary effects of iNO are widely recognized, but the underlying chemistry and pharmacology are poorly understood. Growing evidence suggests
Seung Jin Han et al.
The Journal of endocrinology, 212(3), 307-315 (2011-12-02)
2-Aminobicyclo-(2,2,1)-heptane-2-carboxylic acid (BCH) is an activator of glutamate dehydrogenase (GDH), which is a mitochondrial enzyme with an important role in insulin secretion. We investigated the effect of BCH on the high-glucose (HG)-induced reduction in glucose-stimulated insulin secretion (GSIS), the HG/palmitate
Mikko Gynther et al.
Journal of medicinal chemistry, 51(4), 932-936 (2008-01-26)
The blood-brain barrier efficiently controls the entry of drug molecules into the brain. We describe a feasible means to achieve carrier-mediated drug transport into the rat brain via the specific, large neutral amino acid transporter (LAT1) by conjugating a model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service