22480
(−)-β-Chamigrene
technical, ~90% (sum of enantiomers, GC)
Synonym(s):
(R)-11-Methylene-3,7,7-trimethylspiro[5.5]undec-2-ene
About This Item
grade
technical
assay
~90% (sum of enantiomers, GC)
optical activity
[α]20/D −77±3°, c = 1% in chloroform
refractive index
n20/D 1.511
storage temp.
2-8°C
SMILES string
CC1=CC[C@]2(CC1)C(=C)CCCC2(C)C
InChI
1S/C15H24/c1-12-7-10-15(11-8-12)13(2)6-5-9-14(15,3)4/h7H,2,5-6,8-11H2,1,3-4H3/t15-/m0/s1
InChI key
WLNGPDPILFYWKF-HNNXBMFYSA-N
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service