Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

224669

Sigma-Aldrich

Resolve-Al AgFOD

99%

Synonym(s):

(2,2-Dimethyl-6,6,7,7,8,8,8-heptafluoro-3,5-octanedionato)silver(I), (6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedionato)silver(I), Ag(fod), Silver(I) 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag(OCC(CH3)3CHCOCF2CF2CF3)
CAS Number:
Molecular Weight:
403.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

99%

form

powder

mp

160 °C (dec.) (lit.)

SMILES string

CC(C)(C)\C(O[Ag])=C\C(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10H11F7O2.Ag/c1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h4,18H,1-3H3;/q;+1/p-1/b5-4-;

InChI key

HBFPETJZCPEQFV-MKWAYWHRSA-M

Looking for similar products? Visit Product Comparison Guide

Application

When used in conjunction with a lanthanide shift reagent, provides an efficient NMR shift reagent for aromatic compounds,[1] ammonium salts,[2] and sulfonium and isothiouronium halides.[3]

Legal Information

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 50, 1322-1322 (1985)
Journal of the American Chemical Society, 102, 5903-5903 (1980)
Analytical Chemistry, 59, 562-562 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service