Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

223166

Sigma-Aldrich

Squalene

97%

Synonym(s):

2,6,10,15,19,23-Hexamethyl-2,6,10,14,18,22-tetracosahexaene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2C[=CHCH2CH2C(CH3)]2=CHCH2-]2
CAS Number:
Molecular Weight:
410.72
Beilstein/REAXYS Number:
1728919
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

refractive index

n20/D 1.494 (lit.)

bp

285 °C/25 mmHg (lit.)

mp

−75 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CC(C)=CCCC(C)=CCCC(C)=CCC\C=C(/C)CCC=C(C)CCC=C(C)C

InChI

1S/C30H50/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h15-18,23-24H,9-14,19-22H2,1-8H3/b27-17+,28-18+,29-23+,30-24+

InChI key

YYGNTYWPHWGJRM-AAJYLUCBSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jih-Jung Chen et al.
Journal of natural products, 72(2), 253-258 (2009-02-11)
Five new benzophenone derivatives, 13,14-didehydoxyisogarcinol (1), garcimultiflorone A (2), garcimultiflorone B (3), 13-hydroxygarcimultiflorone B (4), and garcimultiflorone C (5), have been isolated from the fruits of Garcinia multiflora, together with seven known compounds (6-12). The structures of these new compounds
Jeffrey C Noro et al.
Journal of natural products, 71(9), 1623-1624 (2008-08-20)
Tuberculosis has become a major health problem, in particular with the emergence of extremely drug resistant tuberculosis (XDRTB). In our search for new therapeutic leads against TB, we isolated a new triterpene (1) from the plant Rhus taitensis collected in
Tsung-Hsien Chou et al.
Journal of natural products, 73(9), 1470-1475 (2010-08-14)
Bioassay-guided fractionation led to the isolation of six new tetrahydroflavanones, cryptochinones A-F (1-6), from the neutral CHCl(3) fraction of Cryptocarya chinensis leaves, together with 14 known compounds (7-20). The structures of these new compounds were determined through spectroscopic analyses, including
Jih-Jung Chen et al.
Journal of natural products, 72(2), 223-228 (2009-02-06)
Six new compounds, including five new seco-abietane diterpenoids, 12-deoxy-seco-hinokiol methyl ester (1), 12-deoxy-11,12-dihydro-seco-hinokiol methyl ester (2), callicarpic acid A (3), 9alpha-hydroxycallicarpic acid A (4), and callicarpic acid B (5), and a new phenylethanoid derivative, 4-hydroxyphenethyl tetradecanoate (6), have been isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service