Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222976

Sigma-Aldrich

Procaine hydrochloride

99%

Synonym(s):

4-Aminobenzoic acid 2-diethylaminoethyl ester, p-Aminobenzoic acid diethylaminoethyl ester hydrochloride, Novocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2CH2CH2N(C2H5)2·HCl
CAS Number:
Molecular Weight:
272.77
Beilstein/REAXYS Number:
3917802
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

99%

mp

155-156 °C (lit.)

SMILES string

Cl.CCN(CC)CCOC(=O)c1ccc(N)cc1

InChI

1S/C13H20N2O2.ClH/c1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3;1H

InChI key

HCBIBCJNVBAKAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Wiederhold et al.
International immunopharmacology, 29(1), 45-50 (2015-06-30)
We recently identified a specialized cholinergic cell type in tracheal and urethral epithelium that utilizes molecules of the canonical taste transduction signaling cascade to sense potentially harmful substances in the luminal content. Upon stimulation, this cell initiates protective reflexes. Assuming
Jitka Siroká et al.
Journal of pharmaceutical and biomedical analysis, 84, 140-147 (2013-07-09)
A simple capillary electrophoretic method with spectrophotometric UV detection at 236 nm has been developed for the selective separation and determination of 1-(2-chlorophenyl)piperazine (oCPP), 1-(3-chlorophenyl)piperazine (mCPP) and 1-(4-chlorophenyl)piperazine (pCPP) in confiscated pills. Several cyclodextrin derivatives were tested to compose the
Qinlong Liu et al.
Langenbeck's archives of surgery, 397(8), 1323-1331 (2012-11-01)
Ischemia/reperfusion injury is an obstacle especially in steatotic livers, including those with steatosis induced by acute toxic stress. Recently, a modified histidine-tryptophan-ketoglutarate (HTK) solution, HTK-N, has been developed. This solution contains N-acetylhistidine, amino acids, and iron chelators. This study was
E Goldblum et al.
Acta anaesthesiologica Scandinavica, 57(5), 545-552 (2013-01-17)
Spinal anaesthesia is a safe and reliable anaesthetic modality for surgical procedures on the lower part of the body. However, because of the description of transient neurologic symptoms (TNS), most practitioners have abandoned intrathecal lidocaine. Chloroprocaine (2-chloroprocaine, CP) has been
Christopher M Barbieri et al.
Journal of molecular biology, 425(10), 1612-1626 (2013-02-13)
The OmpR/PhoB family of response regulators (RRs) is the largest class of two-component system signal transduction proteins. Extensive biochemical and structural characterization of these transcription factors has provided insights into their activation and DNA-binding mechanisms. For the most part, OmpR/PhoB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service