222917
1,4-Bis(4-methyl-5-phenyl-2-oxazolyl)benzene
suitable for scintillation, 98%
Synonym(s):
1, 4-bis-2-(4-methyl-5-phenyloxazolyl) benzene, Dimethyl-POPOP
About This Item
Recommended Products
assay
98%
mp
233-235 °C (lit.)
suitability
suitable for scintillation
SMILES string
Cc1nc(oc1-c2ccccc2)-c3ccc(cc3)-c4nc(C)c(o4)-c5ccccc5
InChI
1S/C26H20N2O2/c1-17-23(19-9-5-3-6-10-19)29-25(27-17)21-13-15-22(16-14-21)26-28-18(2)24(30-26)20-11-7-4-8-12-20/h3-16H,1-2H3
InChI key
VLDFXDUAENINOO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service