Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

221309

Sigma-Aldrich

Potassium phosphate monobasic

≥99.0%, ACS reagent

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

assay

≥99.0%

impurities

≤0.001% N compounds
≤0.01% insolubles

loss

≤0.2% loss on drying

pH

4.1-4.5

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤0.002%
Na: ≤0.005%
heavy metals: ≤0.001%

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service