Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219932

Sigma-Aldrich

TAPS

Synonym(s):

N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid, [(2-Hydroxy-1,1-bis(hydroxymethyl)ethyl)amino]-1-propanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO6S
CAS Number:
Molecular Weight:
243.28
Beilstein/REAXYS Number:
2452420
EC Number:
MDL number:
Pricing and availability is not currently available.

useful pH range

7.7-9.1

pKa 

8.4

mp

230-235 °C (dec.)

SMILES string

OCC(CO)(CO)NCCCS(O)(=O)=O

InChI

1S/C7H17NO6S/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14/h8-11H,1-6H2,(H,12,13,14)

InChI key

YNLCVAQJIKOXER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Physiological buffer,

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service