Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219886

Sigma-Aldrich

Sodium phosphate dibasic

≥99.0%, ACS reagent

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent

vapor density

4.9 (vs air)

assay

≥99.0%

impurities

≤0.01% insolubles

loss

≤0.2% loss on drying

pH

8.7-9.3

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.002%
heavy metals: ≤0.001%

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for buffer solutions

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro Studies: Inhibition of Nevirapine Metabolism by Nortriptyline in Hepatic Microsomes.
Usach I and PerisJE.
Bio-protocol, 5(19) (2015)
Y C Chang et al.
Molecular and cellular biology, 14(7), 4912-4919 (1994-07-01)
Capsule formation plays a significant role in the pathogenicity of Cryptococcus neoformans. To study the molecular basis of capsule synthesis, the capsule-deficient phenotype of a mutant strain was complemented by transformation. A plasmid rescued from the resulting Cap+ transformant complemented
What is hemoglobin A1c? An analysis of glycated hemoglobins by electrospray ionization mass spectrometry.
Peterson KP, et al.
Clinical Chemistry, 44(9), 1951-1958 (1998)
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have
Natalie K Binder et al.
Molecular human reproduction, 21(5), 424-434 (2015-03-04)
The rate of obesity among men of reproductive age has tripled in the last three decades. Previously, we demonstrated that paternal obesity resulted in impaired preimplantation developmental kinetics, compromised post-compaction metabolism and decreased blastocyst cell number when embryos were generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service