Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21954

Sigma-Aldrich

O-[2-(3-Succinylamino)ethyl]-O′-methyl-polyethylene glycol

PEG average Mn 20,000

Synonym(s):

mono-Methyl polyethylene glycol 20,000 2-(succinylamino)ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

mol wt

PEG average Mn 20,000

Ω-end

carboxylic acid

α-end

methoxy

storage temp.

−20°C

Application

Reactant for:
  • Preparation of supramolecular ABC triblock copolymers via orthogonal self-assembly
  • Filamentous condensation of DNA for transfection studies

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service