Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219312

Sigma-Aldrich

Eserine

99%

Synonym(s):

Physostigmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O2
CAS Number:
Molecular Weight:
275.35
Beilstein/REAXYS Number:
91230
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

99%

optical activity

[α]25/D −116°, c = 1 in benzene

mp

102-104 °C (lit.)

SMILES string

[H][C@]12N(C)CC[C@@]1(C)c3cc(OC(=O)NC)ccc3N2C

InChI

1S/C15H21N3O2/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2/h5-6,9,13H,7-8H2,1-4H3,(H,16,19)/t13-,15+/m1/s1

InChI key

PIJVFDBKTWXHHD-HIFRSBDPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Acetylcholinesterase inhibitor that crosses the blood-brain barrier and forms a carbamylated enzyme complex with acetyl cholinesterase that degrades slowly.
Acetylcholinesterase inhibitor that crosses the blood-brain barrier.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

>212.0 °F

flash_point_c

> 100 °C

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ciaran G Carolan et al.
Journal of medicinal chemistry, 53(3), 1190-1199 (2010-01-14)
Isosorbide-2-benzyl carbamate-5-benzoate is a highly potent and selective BuChE inhibitor. Meanwhile, isosorbide-2-aspirinate-5-salicylate is a highly effective aspirin prodrug that relies on the salicylate portion to interact productively with human BuChE. By integrating the salicylate group into the carbamate design, we
Ciaran G Carolan et al.
Journal of medicinal chemistry, 51(20), 6400-6409 (2008-09-27)
In this study, we report the SAR and characterization of two groups of isosorbide-based cholinesterase inhibitors. The first was based directly on the clinically used nitrate isosorbide mononitrate (ISMN) retention of the 5-nitrate group and introduction of a series of
Weiming Luo et al.
Journal of medicinal chemistry, 49(7), 2174-2185 (2006-03-31)
A new enantiomeric synthesis utilizing classical resolution provided two novel series of optically active inhibitors of cholinesterase: (-)- and (+)-O-carbamoyl phenols of tetrahydrofurobenzofuran and methanobenzodioxepine. An additional two series of (-)- and (+)-O-carbamoyl phenols of pyrroloindole and furoindole were obtained
Weiming Luo et al.
Journal of medicinal chemistry, 48(4), 986-994 (2005-02-18)
Reductive cyclization of 5-hydroxy-3-methyl-3-methoxycarbonylmethylenebenzofuran-2(3H)-one (4) gave 5-hydroxy-3a-methyl-2,3,3a,8a-tatrahydrofuro[2,3-b]benzofuran (5) and the rearrangement product 7-hydroxy-4,5-dihydro-2,5-methano-1,3-benzodioxepine (6). Reaction of compounds 5 and 6 with different isocyanates provided two series novel carbamates (7-12) whose structures were confirmed by X-ray crystallography. These were assessed for
Federica Belluti et al.
Journal of medicinal chemistry, 48(13), 4444-4456 (2005-06-25)
In continuing research that led us to identify a new class of carbamate derivatives acting as potent (Rampa et al. J. Med. Chem. 1998, 41, 3976) and long-lasting (Rampa et al. J. Med. Chem. 2001, 44, 3810) acetylcholinesterase (AChE) inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service