Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219134

Sigma-Aldrich

Betaine monohydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2COO- · H2O
CAS Number:
Molecular Weight:
135.16
Beilstein/REAXYS Number:
3626258
EC Number:
MDL number:

assay

99%

SMILES string

O.C[N+](C)(C)CC([O-])=O

InChI

1S/C5H11NO2.H2O/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H2

InChI key

NJZRLXNBGZBREL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

End-product of oxidative metabolism of choline, betaine is a general methyl donor, in particular in a minor pathway of methionine biosynthesis. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service