Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217824

Sigma-Aldrich

Adipic acid dihydrazide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NHCO(CH2)4CONHNH2
CAS Number:
Molecular Weight:
174.20
Beilstein/REAXYS Number:
973863
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

180-182 °C (lit.)

SMILES string

NNC(=O)CCCCC(=O)NN

InChI

1S/C6H14N4O2/c7-9-5(11)3-1-2-4-6(12)10-8/h1-4,7-8H2,(H,9,11)(H,10,12)

InChI key

IBVAQQYNSHJXBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anupam Basu et al.
Clinica chimica acta; international journal of clinical chemistry, 366(1-2), 287-292 (2005-12-13)
In hapten enzyme immunoassays (EIA), there is an increase or decrease of labeled hapten recognition by antibody that affects sensitivity of the assay. We incorporated a spacer between a hapten derivative and enzyme to test its influence on the sensitivity
P Brne et al.
Journal of chromatography. A, 1216(13), 2658-2663 (2009-02-11)
Convective interaction media (CIM; BIA Separations) monoliths are attractive stationary phases for use in affinity chromatography because they enable fast affinity binding, which is a consequence of convectively enhanced mass transport. This work focuses on the development of novel CIM
Wen-Yu Su et al.
Acta biomaterialia, 6(8), 3044-3055 (2010-03-03)
Injectable hydrogel allows irregular surgical defects to be completely filled, lessens the risk of implant migration, and minimizes surgical defects due to the solution-gel state transformation. Here, we first propose a method for preparing oxidized hyaluronic acid/adipic acid dihydrazide (oxi-HA/ADH)
Tulsidas G Shrivastav
Journal of immunoassay & immunochemistry, 24(3), 301-309 (2003-09-05)
Preparation of horseradish peroxidase (HRP) hydrazide that is HRP linked to adipic acid dihydrazide (HRP-ADH) and its use in enzyme immunoassay (EIA) is described. In this new strategy, horseradish peroxidase was conjugated to adipic acid dihydrazide using a carbodiimide coupling
Wen-Yu Su et al.
Journal of biomaterials science. Polymer edition, 22(13), 1777-1797 (2010-09-17)
Vitrectomy is a common procedure for treating ocular-related diseases. The surgery involves removing the vitreous humor from the center of the eye, and vitreous substitutes are needed to replace the vitreous humor after vitrectomy. In the present study, we developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service