Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217131

Sigma-Aldrich

Decahydronaphthalene-d18

98 atom % D

Synonym(s):

Decalin-d18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D18
CAS Number:
Molecular Weight:
156.36
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

description

cis/trans mixture

isotopic purity

98 atom % D

assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.475 (lit.)

bp

193 °C (lit.)

density

1.012 g/mL at 25 °C

mass shift

M+18

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])C2([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C2([2H])C1([2H])[2H]

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D,10D

InChI key

NNBZCPXTIHJBJL-XRRAJICISA-N

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

Decalin is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

135.0 °F - closed cup

flash_point_c

57.20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of 4-and 4, 5-substituted 2, 7-di-tert-butylthiepins.
Yamamoto K, et al.
Chemistry Letters (Jpn), 11, 1843-1846 (1982)
THE PROTON MAGNETIC RESONANCE SPECTRUM OF ETHYLENIMINE.
Nakanishi H and Yamamoto O.
Chemistry Letters (Jpn), 5, 465-468 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service