Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

214132

Sigma-Aldrich

4,5-Dihydro-3-(4-pyridinyl)-2H-benz[g]indazole,methanesulfonate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3O3S
CAS Number:
Molecular Weight:
343.40
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

95%

mp

275 °C (subl.) (lit.)

SMILES string

CS(O)(=O)=O.C1Cc2c(n[nH]c2-c3ccncc3)-c4ccccc14


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service