Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

212385

Sigma-Aldrich

Direct Violet 51

Dye content 50 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H27N5Na2O8S2
CAS Number:
Molecular Weight:
719.70
Colour Index Number:
27905
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 50%

λmax

549 nm

SMILES string

[Na+].[Na+].COc1cc(N=Nc2c(C)cc(C)cc2S([O-])(=O)=O)c(C)cc1N=Nc3c(O)c4ccc(Nc5ccccc5)cc4cc3S([O-])(=O)=O

InChI

1S/C32H29N5O8S2.2Na/c1-18-12-20(3)30(28(13-18)46(39,40)41)36-34-25-17-27(45-4)26(14-19(25)2)35-37-31-29(47(42,43)44)16-21-15-23(10-11-24(21)32(31)38)33-22-8-6-5-7-9-22;;/h5-17,33,38H,1-4H3,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2

InChI key

LARMRMCFZNGNNX-UHFFFAOYSA-L

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunichi Asai et al.
FEBS letters, 597(8), 1164-1175 (2023-02-09)
Cholinergic receptor muscarinic 3 (CHRM3)-mediated focal adhesion kinase/YES-associated protein (YAP) signalling is essential for the growth of castration-resistant prostate cancer (CRPC) cells. Here, we evaluated the molecular mechanisms through which CHRM3 overexpression facilitates castration-resistant growth. Small RNA sequencing combined with
Richard T Waldron et al.
The Journal of physiology, 597(12), 3085-3105 (2019-05-06)
This work confirms previous reports that CM4620, a small molecule inhibitor of Ca2+ entry via store operated Ca2+ entry (SOCE) channels formed by stromal interaction molecule 1 (STIM1)/Orai complexes, attenuates acinar cell pathology and acute pancreatitis in mouse experimental models.
Killian Chaumonnot et al.
Cell death & disease, 12(1), 114-114 (2021-01-24)
The role of GRP94, an endoplasmic reticulum (ER) stress protein with both pro- and anti-inflammatory functions, has not been investigated in macrophages during ER stress, whereas ER stress has been reported in many diseases involving macrophages. In this work, we
Wenjie Yu et al.
Developmental cell, 57(18), 2221-2236 (2022-09-16)
Pulmonary neuroendocrine cells (PNECs) are rare airway cells with potential sensory capacity linked to vagal neurons and immune cells. How PNECs sense and respond to external stimuli remains poorly understood. We discovered PNECs located within pig and human submucosal glands
Scott M Emrich et al.
Cell reports, 34(9), 108760-108760 (2021-03-04)
Stromal-interaction molecules (STIM1/2) sense endoplasmic reticulum (ER) Ca2+ depletion and activate Orai channels. However, the choreography of interactions between native STIM/Orai proteins under physiological agonist stimulation is unknown. We show that the five STIM1/2 and Orai1/2/3 proteins are non-redundant and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service