Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

211397

Sigma-Aldrich

2-Methylglutaronitrile

technical grade, 80%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH(CH3)CH2CH2CN
CAS Number:
Molecular Weight:
108.14
Beilstein/REAXYS Number:
1741955
MDL number:

grade

technical grade

assay

80%

impurities

≤0.5% benzene

refractive index

n20/D 1.434 (lit.)

bp

269-271 °C (lit.)

mp

−45 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CC(CCC#N)C#N

InChI

1S/C6H8N2/c1-6(5-8)3-2-4-7/h6H,2-3H2,1H3

InChI key

FPPLREPCQJZDAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service