Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

210668

Sigma-Aldrich

Acid Violet 7

Dye content 40 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N4Na2O9S2
CAS Number:
Molecular Weight:
566.47
Colour Index Number:
18055
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 40%

λmax

520 nm

SMILES string

[Na+].[Na+].CC(=O)Nc1ccc(cc1)\N=N\c2c(O)c3c(NC(C)=O)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C20H18N4O9S2.2Na/c1-10(25)21-13-3-5-14(6-4-13)23-24-19-17(35(31,32)33)8-12-7-15(34(28,29)30)9-16(22-11(2)26)18(12)20(19)27;;/h3-9,27H,1-2H3,(H,21,25)(H,22,26)(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2/b24-23+;;

InChI key

HRMOLDWRTCFZRP-KPOOZVEVSA-L

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guang Guo et al.
Chemosphere, 244, 125510-125510 (2019-12-15)
As a result of the use of a large amount of salt in dye industries, azo dye decolorization is often needed under hypersaline environments and low dissolved oxygen. Consortium GG-1, which is able to decolorize azo dyes in high salt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service