Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

210552

Sigma-Aldrich

Acid Blue 41

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N3NaO6S
CAS Number:
Molecular Weight:
489.48
Colour Index Number:
62130
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 90%

SMILES string

[Na+].CN(C(C)=O)c1ccc(Nc2cc(c(N)c3C(=O)c4ccccc4C(=O)c23)S([O-])(=O)=O)cc1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service