Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21010

Sigma-Aldrich

Cesium nitrate

purum p.a., ≥99.0% (T)

Synonym(s):

Nitric acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsNO3
CAS Number:
Molecular Weight:
194.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum p.a.

assay

≥99.0% (T)

mp

414 °C (lit.)

density

3.685 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤300 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Cs+].[O-]N(=O)=O

InChI

1S/Cs.NO3/c;2-1(3)4/q+1;-1

InChI key

NLSCHDZTHVNDCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of cesium nitratocuprate, Cs2[Cu(NO3)4], which crystallizes in a square-planar coordination around the Cu center.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1 - Repr. 2

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service