Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208426

Sigma-Aldrich

Sodium perchlorate

98%

Synonym(s):

Hyperchloric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO4
CAS Number:
Molecular Weight:
122.44
EC Number:
MDL number:
UNSPSC Code:
12352302

assay

98%

mp

468 °C (lit.)

SMILES string

[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

BAZAXWOYCMUHIX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1 - STOT RE 2

target_organs

Thyroid

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Cincinelli et al.
Bioorganic & medicinal chemistry, 19(6), 1847-1851 (2011-03-08)
A high activity α-carbonic anhydrase (CA, EC 4.2.1.1) has been purified from various tissues of the Antarctic seal Leptonychotes weddellii. The new enzyme, denominated lwCA, has a catalytic activity for the physiologic CO(2) hydration to bicarbonate reaction, similar to that
Peter Burghout et al.
Bioorganic & medicinal chemistry, 19(1), 243-248 (2010-12-18)
The Gram-positive bacterium Streptococcus pneumoniae is a human respiratory tract pathogen that contributes significantly to global mortality and morbidity. It was recently shown that this bacterial pathogen depends on a conserved β-carbonic anhydrase (CA, EC 4.2.1.1) for in vitro growth
Daniela Vullo et al.
Bioorganic & medicinal chemistry letters, 21(12), 3591-3595 (2011-05-17)
Two new β-carbonic anhydrases (CAs, EC 4.2.1.1) from the bacterial pathogen Salmonella enterica serovar Typhimurium, stCA 1 and stCA 2, were characterized kinetically. The two enzymes possess appreciable activity as catalysts for the hydration of CO(2) to bicarbonate, with k(cat)
Anna Ohradanova et al.
Bioorganic & medicinal chemistry letters, 22(3), 1314-1316 (2012-01-10)
An α-carbonic anhydrase (CA, EC 4.2.1.1) isolated from the living fossil sponge Astrosclera willeyana, Astrosclerin, was investigated for its inhibition profile with simple inorganic anions, complex anions and other small molecules known to interact with these zinc enzymes. Astrosclerin is
Simran Kaur et al.
Journal of food protection, 78(9), 1708-1718 (2015-09-01)
Previous studies show that treatment of cantaloupes with chlorine dioxide (ClO2) gas at 5 mg/liter for 10 min results in a significant reduction (P < 0.05) in initial microflora, an increase in shelf life without any alteration in color, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service