Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208132

Sigma-Aldrich

Mercury(II) thiocyanate

98%

Synonym(s):

Mercuric isothiocyanate, Mercuric sulfocyanide, Mercuric thiocyanate, Mercury di(thiocyanate), Thiocyanic acid, mercury(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(SCN)2
CAS Number:
Molecular Weight:
316.75
Beilstein/REAXYS Number:
3561344
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

98%

mp

165 °C (dec.) (lit.)

density

3.71 g/mL at 25 °C (lit.)

SMILES string

N#CS[Hg]SC#N

InChI

1S/2CHNS.Hg/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

GBZANUMDJPCQHY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

application

Increasing interest in solid state NMR of heavy-metal nuclei results in the preparation of several Hg-thiocyanate and selenocyanate compounds as potential models.[1]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bowmaker, G.A. et al.
Inorganic Chemistry, 37, 1734-1734 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service