Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

207756

Sigma-Aldrich

N-[2-(Diethylamino)ethyl]-4-ethoxybenzamide hydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2 · HCl
CAS Number:
Molecular Weight:
300.82
MDL number:
Pricing and availability is not currently available.

assay

99%

SMILES string

Cl.CCOc1ccc(cc1)C(=O)NCCN(CC)CC

InChI

1S/C15H24N2O2.ClH/c1-4-17(5-2)12-11-16-15(18)13-7-9-14(10-8-13)19-6-3;/h7-10H,4-6,11-12H2,1-3H3,(H,16,18);1H

InChI key

DBBNMTPWROZAFK-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service