Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

206148

Sigma-Aldrich

Rhenium on alumina

extent of labeling: 0.5 wt. % loading, matrix alumina (unreduced), pellets, 3.2 mm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

pellets

extent of labeling

0.5 wt. % loading

matrix

alumina (unreduced)

particle size

3.2 mm

SMILES string

[ReH]

General description

unreduced

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service