Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

205443

Sigma-Aldrich

4-Ethylbenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4CH2OH
CAS Number:
Molecular Weight:
136.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.527 (lit.)

bp

115-117 °C/9 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(CO)cc1

InChI

1S/C9H12O/c1-2-8-3-5-9(7-10)6-4-8/h3-6,10H,2,7H2,1H3

InChI key

YSLBFFIVJGJBSA-UHFFFAOYSA-N

Application

4-Ethylbenzyl alcohol may be used in chemical synthesis studies.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annemieke Strijkstra et al.
Applied and environmental microbiology, 80(24), 7592-7603 (2014-09-28)
The betaproteobacteria "Aromatoleum aromaticum" pCyN1 and "Thauera" sp. strain pCyN2 anaerobically degrade the plant-derived aromatic hydrocarbon p-cymene (4-isopropyltoluene) under nitrate-reducing conditions. Metabolite analysis of p-cymene-adapted "A. aromaticum" pCyN1 cells demonstrated the specific formation of 4-isopropylbenzyl alcohol and 4-isopropylbenzaldehyde, whereas with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service