Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201952

Sigma-Aldrich

Tartrazine

Dye content 80 %

Synonym(s):

Acid Yellow 23

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9N4Na3O9S2
CAS Number:
Molecular Weight:
534.36
Colour Index Number:
19140
Beilstein/REAXYS Number:
69850
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 80%

λmax

425 nm

SMILES string

[Na+].[Na+].[Na+].[O-]C(=O)C1=NN(c2ccc(cc2)S([O-])(=O)=O)C(=O)C1N=Nc3ccc(cc3)S([O-])(=O)=O

InChI

1S/C16H12N4O9S2.3Na/c21-15-13(18-17-9-1-5-11(6-2-9)30(24,25)26)14(16(22)23)19-20(15)10-3-7-12(8-4-10)31(27,28)29;;;/h1-8,13H,(H,22,23)(H,24,25,26)(H,27,28,29);;;/q;3*+1/p-3

InChI key

UJMBCXLDXJUMFB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service