Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201642

Sigma-Aldrich

Fluorescein diacetate

98%

Synonym(s):

3,6-Diacetoxyfluoran, Di-O-acetylfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16O7
CAS Number:
Molecular Weight:
416.38
Beilstein/REAXYS Number:
62575
EC Number:
MDL number:
UNSPSC Code:
12171500

assay

98%

mp

200-203 °C (lit.)

λmax

498 nm

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C24H16O7/c1-13(25)28-15-7-9-19-21(11-15)30-22-12-16(29-14(2)26)8-10-20(22)24(19)18-6-4-3-5-17(18)23(27)31-24/h3-12H,1-2H3

InChI key

CHADEQDQBURGHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David C Gillan et al.
The Science of the total environment, 419, 98-108 (2012-01-28)
Muddy sediments of the Belgian Continental Zone (BCZ) are contaminated by metals such as Co, As, Cd, Pb, and Ni. Previous studies have suggested that mineralization of phytodetritus accumulating each year on sediments might cause secondary contaminations of the overlying
Melina M Malinen et al.
Tissue engineering. Part A, 18(23-24), 2418-2425 (2012-06-21)
Current hepatocyte models do not mimic the human liver morphology and functions properly and, therefore, drug metabolism, excretion, and toxicity in the liver are inadequately predicted. In this study, we established three-dimensional (3D) hepatic cell cultures in hydrogels of peptide
Koichi Baba et al.
Journal of controlled release : official journal of the Controlled Release Society, 153(3), 278-287 (2011-05-10)
This report describes a method for enhancing the ocular penetration of eye drops using nanoparticles of hydrolyzable dye, which is similar to a prodrug approach. The entry of eye drops into the ocular globe is restricted predominantly by corneal barrier
Design of multifunctional micelle for tumor-targeted intracellular drug release and fluorescent imaging.
Weiwei Wang et al.
Advanced materials (Deerfield Beach, Fla.), 24(1), 115-120 (2011-12-07)
Nicole Birgit Arweiler et al.
Clinical oral investigations, 16(2), 499-504 (2011-03-02)
Preliminary data have suggested that taurolidine may bear promising disinfectant properties for the therapy of bacterial infections. However, at present, the potential antibacterial effect of taurolidine on the supragingival plaque biofilm is unknown. To evaluate the antibacterial effect of taurolidine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service