Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

201111

Sigma-Aldrich

Antimony(III) fluoride

98%

Synonym(s):

Antimony trifluoride, Trifluorostibine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbF3
CAS Number:
Molecular Weight:
178.76
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

98%

reaction suitability

reagent type: catalyst
core: antimony

mp

235 °C (dec.) (lit.)

density

4.38 g/mL at 25 °C (lit.)

SMILES string

F[Sb](F)F

InChI

1S/3FH.Sb/h3*1H;/q;;;+3/p-3

InChI key

GUNJVIDCYZYFGV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service