Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200980

Sigma-Aldrich

5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6Cl4O2
CAS Number:
Molecular Weight:
263.93
Beilstein/REAXYS Number:
477013
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.525

bp

108-110 °C/11 mmHg (lit.)

density

1.501 g/mL at 25 °C (lit.)

SMILES string

COC1(OC)C(Cl)=C(Cl)C(Cl)=C1Cl

InChI

1S/C7H6Cl4O2/c1-12-7(13-2)5(10)3(8)4(9)6(7)11/h1-2H3

InChI key

UHSMEJQTFMHABA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene forms endo-adduct with 1,3-cyclopentadiene[1].

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polycyclitols: synthesis of novel carbasugar and conduritol analogues as potential glycosidase inhibitors.
Tetrahedron Letters, 42(10), 1987-1990 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service