19908
4-tert-Butyldimethylsiloxy-3-penten-2-one
≥97.0% (GC)
Synonym(s):
Acetylacetone enol tert-butyldimethylsilyl ether
About This Item
assay
≥97.0% (GC)
refractive index
n20/D 1.462
bp
48-50 °C/0.2 mmHg
density
0.907 g/mL at 20 °C (lit.)
SMILES string
CC(=O)\C=C(/C)O[Si](C)(C)C(C)(C)C
InChI
1S/C11H22O2Si/c1-9(12)8-10(2)13-14(6,7)11(3,4)5/h8H,1-7H3/b10-8+
InChI key
ZRVOCNOYAJIAAP-CSKARUKUSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
176.0 °F - closed cup
flash_point_c
80 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service