Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

198331

Sigma-Aldrich

Cesium hydroxide monohydrate

Synonym(s):

2-Aminoethyl 2-methylacrylate hydrochloride, 2-Aminoethyl methacrylate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCsO · H2O
CAS Number:
Molecular Weight:
167.93
EC Number:
MDL number:
UNSPSC Code:
12352300

impurities

<5% Cs2CO3

mp

272 °C (lit.)

density

3.68 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[O-][H].[H]O[H]

InChI

1S/Cs.H2O/h;1H2/q+1;/p-1

InChI key

HUCVOHYBFXVBRW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Johannes Scholz et al.
Magma (New York, N.Y.), 28(6), 591-598 (2015-10-10)
(13)C metabolic MRI using hyperpolarized (13)C-bicarbonate enables preclinical detection of pH. To improve signal-to-noise ratio, experimental procedures were refined, and the influence of pH, buffer capacity, temperature, and field strength were investigated. Bicarbonate preparation was investigated. Bicarbonate was prepared and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service