Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197939

Sigma-Aldrich

Borane tert-butylamine complex

97%, pellets, diam. 11 mm

Synonym(s):

tert-Butylamine borane, NSC 114045, Trihydro(2-methyl-2-propanamine) boron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNH2 · BH3
CAS Number:
Molecular Weight:
86.97
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

97%

form

pellets

reaction suitability

reagent type: reductant

diam.

11 mm

weight

~0.4 g (pellet)

mp

98-100 °C (dec.) (lit.)

SMILES string

B.CC(C)(C)N

InChI

1S/C4H11N.BH3/c1-4(2,3)5;/h5H2,1-3H3;1H3

InChI key

GKFJEDWZQZKYHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A water-soluble borane reducing agent
Reactant involved in:
  • Non-destructive analyses and conservation treatments of Indian drawings
  • Dehydrogenation reactions mediated by ruthenium bidentate phosphine complexes or ruthenium / iridium bis(N-heterocyclic carbene) complexes
  • The formation of σ-borane complexes
  • Ethanolysis of amine-borane adducts to form a reducing system for transfer hydrogenation of terminal olefins
  • Photochemical hydroboration and oxidation of single-walled carbon nanotubes

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grant E Johnson et al.
Physical chemistry chemical physics : PCCP, 17(22), 14636-14646 (2015-05-15)
We present a systematic study of the effect of the number of methyl (Me) and cyclohexyl (Cy) functional groups in monodentate phosphine ligands on the solution-phase synthesis of ligated sub-nanometer gold clusters and their gas-phase fragmentation pathways. Small mixed ligand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service