Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

196630

Sigma-Aldrich

2,2′,3,3′,5,5′,6,6′-Octafluorobiphenyl

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F4C6F4H
CAS Number:
Molecular Weight:
298.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

solid

mp

84-86 °C (lit.)

SMILES string

Fc1cc(F)c(F)c(c1F)-c2c(F)c(F)cc(F)c2F

InChI

1S/C12H2F8/c13-3-1-4(14)10(18)7(9(3)17)8-11(19)5(15)2-6(16)12(8)20/h1-2H

InChI key

QWCHHUZAAGRHDB-UHFFFAOYSA-N

Application

2,2′,3,3′,5,5′,6,6′-Octafluorobiphenyl was used in the preparation of nickel octafluorobiphenyl complexes[1].

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallic Polymer and Linear Mono-, Bi-, and Trimetailk Octafluoro-p, p'-blphenylene-Bridged Complexes of Bls (methyldiphenylphosphine) nickel: X-ray Crystal Structures of NI (PMePh2),(4, 4'-C12F, H) Br and Ni (PMePh2),(4, 4 '-C12F8H) 21i2.
Sturge KC, et al.
Organometallics, 11(9), 3056-3062 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service