Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

192449

Sigma-Aldrich

4,4′-Isopropylidenebis(2,6-dichlorophenol)

98%

Synonym(s):

2,2′,6,6′-Tetrachloro-4,4′-isopropylidenediphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C[C6H2(Cl)2OH]2
CAS Number:
Molecular Weight:
366.07
EC Number:
MDL number:
UNSPSC Code:
12162002

assay

98%

mp

134-136 °C (lit.)

SMILES string

CC(C)(c1cc(Cl)c(O)c(Cl)c1)c2cc(Cl)c(O)c(Cl)c2

InChI

1S/C15H12Cl4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3

InChI key

KYPYTERUKNKOLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Sun et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 44(11), 1916-1921 (2006-08-09)
Previously, we developed an androgen receptor (AR)-mediated reporter gene assay system, and verified the antiandrogenic activity of bisphenol A (BPA) based on this assay. There were some similar phenol-ring structure chemicals which were widely used while little had been done
M F Fernandez et al.
Reproductive toxicology (Elmsford, N.Y.), 24(2), 259-264 (2007-08-11)
Bisphenol-A (BPA) and chlorinated derivatives (Cl(x)BPA) were investigated in adipose tissue of women in Southeast Spain. BPA was above limit of detection (LOD) in 11 out of 20 samples (55%). Among Cl(x)BPA, Cl(2)BPA was the most frequent (80%) and abundant
Patricia Moreira Bastos et al.
Chemosphere, 70(7), 1196-1202 (2007-09-28)
The term persistence has been used to confusion since it is used as a conceptual parameter without a uniform definition. Work is therefore being done in order to unite ideas and describe persistence based on the chemical reactivity and chemico-physical
Shigeyuki Kitamura et al.
Biochemical and biophysical research communications, 293(1), 554-559 (2002-06-11)
The thyroid hormonal-disrupting activity of the flame retardants tetrabromobisphenol A (TBBPA) and tetrachlorobisphenol A (TCBPA) was examined and compared with that of bisphenol A, a typical estrogenic xenobiotic. TBBPA and TCBPA, halogenated derivatives of bisphenol A, markedly inhibited the binding
Consuelo Sánchez-Brunete et al.
Journal of chromatography. A, 1216(29), 5497-5503 (2009-06-16)
In this work, an isotope dilution method for the determination, in agricultural and industrial soil samples, of tetrabromobisphenol-A, tetrachlorobisphenol-A and bisphenol-A by gas chromatography-mass spectrometry was developed. The compounds were extracted from soil by sonication assisted extraction in small columns

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service