Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

191736

Sigma-Aldrich

Pargyline hydrochloride

99%

Synonym(s):

N-Methyl-N-(2-propynyl)benzylamine hydrochloride, N-Methyl-N-propargylbenzylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)CH2C≡CH·HCl
CAS Number:
Molecular Weight:
195.69
Beilstein/REAXYS Number:
3699487
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

99%

mp

160-163 °C (lit.)

SMILES string

Cl[H].CN(CC#C)Cc1ccccc1

InChI

1S/C11H13N.ClH/c1-3-9-12(2)10-11-7-5-4-6-8-11;/h1,4-8H,9-10H2,2H3;1H

Inchi Key

BCXCABRDBBWWGY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective MAO-B inhibitor.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Reineri et al.
Journal of the American Chemical Society, 134(27), 11146-11152 (2012-06-06)
(15)N-Propargylcholine has been synthesized and hydrogenated with para-H(2). Through the application of a field cycling procedure, parahydrogen spin order is transferred to the (15)N resonance. Among the different isomers formed upon hydrogenation of (15)N-propargylcholine, only the nontransposed derivative contributes to
Masahiro Yoshida et al.
The Journal of organic chemistry, 78(4), 1687-1692 (2013-02-01)
The reaction of propargylic carbonates with 4-hydroxy-2-pyrones in the presence of a palladium catalyst is described. By the choice of the reaction temperature, two types of the substituted furo[3,2-c]pyran-4-one derivatives were regioselectively synthesized, respectively.
Lavinia Boccia et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(11), 15448-15461 (2020-09-29)
The LPBN (lateral parabrachial nucleus) plays an important role in feeding control. CGRP (calcitonin gene-related peptide) LPBN neurons activation mediates the anorectic effects of different gut-derived peptides, including amylin. Amylin and its long acting analog sCT (salmon calcitonin) exert their
Shauna N Vasilatos et al.
Carcinogenesis, 34(6), 1196-1207 (2013-01-29)
Our previous studies demonstrated that lysine-specific demethylase 1 (LSD1) and histone deacetylases (HDACs) closely interact in controlling growth of breast cancer cells. However, the underlying mechanisms are largely unknown. In this study, we showed that knockdown of LSD1 expression (LSD1-KD)
Zheng Jiang et al.
Organic letters, 14(24), 6266-6269 (2012-12-01)
An efficient and straightforward strategy for the synthesis of tetrasubstituted imidazoles from propargyl amines, sulfonyl azides, and terminal alkynes is described. N-sulfonyl ketenimine and aminoallene are believed to be the key intermediates for this two-step one-pot transformation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service