Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

191132

Sigma-Aldrich

2-Methyl-1-butanethiol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH2SH
CAS Number:
Molecular Weight:
104.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

41.4 mmHg ( 37.7 °C)

assay

95%

refractive index

n20/D 1.447 (lit.)

bp

116-117 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

SMILES string

CCC(C)CS

InChI

1S/C5H12S/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3

InChI key

WGQKBCSACFQGQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

66.2 °F - closed cup

flash_point_c

19 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service