Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

185795

Sigma-Aldrich

Cumene

99%

Synonym(s):

(1-Methylethyl)benzene, 2-Phenylpropane, Isopropylbenzene, NSC 8776

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)2
CAS Number:
Molecular Weight:
120.19
Beilstein/REAXYS Number:
1236613
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

4.1 (vs air)

vapor pressure

8 mmHg ( 20 °C)
9.7 mmHg ( 37.7 °C)

assay

99%

autoignition temp.

797 °F

expl. lim.

6.5 %

refractive index

n20/D 1.491 (lit.)

bp

152-154 °C (lit.)

mp

−96 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccccc1

InChI

1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

RWGFKTVRMDUZSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

87.8 °F - closed cup

flash_point_c

31.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Bråred Christensson et al.
Contact dermatitis, 55(4), 230-237 (2006-09-09)
Concomitant positive reactions to colophonium, oxidized limonene, and/or oxidized linalool are recorded in patch test studies. The main allergens in these patch test mixtures are hydroperoxides, which form antigens by a radical pathway. Theoretically, concomitant reactions can be explained not
Mathieu Frenette et al.
Journal of the American Chemical Society, 128(51), 16432-16433 (2006-12-21)
A new class of thermally activated chain-breaking antioxidants is presented. Dimers of persistent carbon-centered radicals are able to inhibit the autoxidation of cumene and styrene with better rate constants than the commercial antioxidant Irganox HP-136 and 3,5-di-tert-butyl-4-hydroxyanisole. A dramatic increase
A Getsoian et al.
International journal of pharmaceutics, 348(1-2), 3-9 (2007-10-20)
To emphasize the fact that solvents can be either critical or immaterial in crystallizing specific polymorphs, a method for obtaining multiple polymorphs of a compound using only one solvent is demonstrated. By varying the crystallization temperature and level of supersaturation
So-Young Jun et al.
Biochimica et biophysica acta, 1764(7), 1159-1166 (2006-07-18)
The meta-cleavage product hydrolase from Pseudomonas fluorescens IP01 (CumD) hydrolyzes 2-hydroxy-6-oxo-7-methylocta-2,4-dienoate (6-isopropyl HODA) in the cumene (isopropylbenzene) degradation pathway. To modulate the substrate specificity and catalytic efficiency of CumD toward substrates derived from monocyclic aromatic compounds, we constructed the CumD
Youji Okada et al.
Organic & biomolecular chemistry, 6(6), 1097-1102 (2008-03-11)
The antioxidant effects of the new thiosulfinate derivative, S-benzyl phenylmethanethiosulfinate (BPT), against the oxidation of cumene and methyl linoleate (ML) in chlorobenzene were studied in detail using HPLC. The results showed that BPT provided effective inhibition with a well-defined induction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service