Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

184896

Sigma-Aldrich

2-Methylnaphthalene-bis(hexachlorocyclopentadiene) adduct

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H10Cl12
CAS Number:
Molecular Weight:
687.74
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

158-161 °C (lit.)

SMILES string

Cc1ccc2C3C(C4C(c2c1)[C@]5(Cl)C(Cl)=C(Cl)[C@@]4(Cl)C5(Cl)Cl)[C@]6(Cl)C(Cl)=C(Cl)[C@@]3(Cl)C6(Cl)Cl

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to substituted naphthalenes which are otherwise difficult to synthesize.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 7, 23-23 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service