Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

184446

Sigma-Aldrich

3-Pyridylcarbinol N-oxide

97%

Synonym(s):

3-Pyridinemethanol N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO2
CAS Number:
Molecular Weight:
125.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

87-89 °C (lit.)

SMILES string

OCc1ccc[n+]([O-])c1

InChI

1S/C6H7NO2/c8-5-6-2-1-3-7(9)4-6/h1-4,8H,5H2

Inchi Key

LQYJAVWWKTWWKN-UHFFFAOYSA-N

Application

3-Pyridylcarbinol N-oxide was used to develop a series of urinary biomarkers to assess the metabolic activation and detoxification of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone in humans[1].

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S G Carmella et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 6(2), 113-120 (1997-02-01)
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) is a potent pulmonary carcinogen in rodents and is believed to be a causative factor for lung cancer in smokers. NNK also may be involved in oral cancer etiology in users of smokeless tobacco products. Pyridine-N-oxidation of NNK

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service