Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18225

Sigma-Aldrich

4-(2-Bromopropionyl)phenoxyacetic acid

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11BrO4
CAS Number:
Molecular Weight:
287.11
Beilstein/REAXYS Number:
2376939
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.0%

reaction suitability

reagent type: cross-linking reagent

mp

139-142 °C

SMILES string

CC(Br)C(=O)c1ccc(OCC(O)=O)cc1

InChI

1S/C11H11BrO4/c1-7(12)11(15)8-2-4-9(5-3-8)16-6-10(13)14/h2-5,7H,6H2,1H3,(H,13,14)

InChI key

SQLRCYTWVMDUBW-UHFFFAOYSA-N

Other Notes

Photolabile linker for solid-phase peptide synthesis[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Belloff et al.
Chimia, 39, 317-317 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service