Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

181951

Sigma-Aldrich

Polyethylene, chlorosulfonated

average Mw ~195,000 by GPC, 1.1 wt. % S (as SO2Cl)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002

form

chunks

mol wt

average Mw ~195,000 by GPC

concentration

1.1 wt. % S (as SO2Cl)

extent of labeling

43 wt. % Cl


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service